Learn More
HLA-B7 Monoclonal Antibody (BB7.1), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand:  Affymetrix eBioscience 46-5917-41
 View more versions of this product
Product Code. 16370864
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Mouse Monoclonal Antibody
HLA-B7 Monoclonal antibody specifically detects HLA-B7 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HLA-B7
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P01889
HLA-B
Papain solubilized HLA-B7
25 Tests
Primary
Human
Antibody
IgG1 κ
Flow Cytometry
BB7.1
PerCP-eFluor 710
HLA-B
AS; Aw-33; Aw-74; B-4901; B-5001; Bw-47; Bw-50; CDABP0067; DAQB-90C11.16; FLJ26655; HEL-S-83; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-3 alpha chain; HLA class I histocompatibility antigen, B-7 alpha chain; HLAA; HLA-A; HLA-A11; HLA-A33; HLAB; HLA-B; HLA-B 1502; HLA-B*45ZJ; HLA-B15; HLA-B-3506; HLA-B39; HLA-B-3905; HLA-B49; HLA-B50; HLA-B55; HLA-B-5502; HLA-B-5602; HLA-B59; HLA-B61; HLA-Cw; HLA-DQB1; HLA-DRB1; MHC 1; MHC class I antigen A*1; MHC class I antigen A*3; MHC class I antigen B*7; MHC1; SPDA1
Mouse
Affinity chromatography
RUO
3106
4° C, store in dark, DO NOT FREEZE!