missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-B Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA5143261
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:64,000.
HLA-B belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. Class I molecules play a central role in the immune system by presenting peptides derived from the endoplasmic reticulum lumen. They are expressed in nearly all cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon 1 encodes the leader peptide, exon 2 and 3 encode the alpha1 and alpha2 domains, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region and exons 6 and 7 encode the cytoplasmic tail. Polymorphisms within exon 2 and exon 3 are responsible for the peptide binding specificity of each class one molecule. Typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. Hundreds of HLA-B alleles have been described. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Specifications
HLA-B | |
Polyclonal | |
Unconjugated | |
HLA-B | |
A29 protein; antigen presenting molecule; AS; Aw-33; Aw-74; B-4901; B-5001; Bw-47; Bw-50; class I MHC, HLA-B antigen; DAQB-90C11.16; epididymis secretory protein Li 83; FLJ26655; HEL-S-83; histocompatibility antigen; HLA A; HLA -B; HLA B-40011; HLA B44; HLA class I antigen; HLA class I antigen HLA-B; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA class I histocompatibility antigen, B alpha chain; HLA class I histocompatibility antigen, B-47 alpha chain; HLA class I histocompatibility antigen, B-50 alpha chain; HLA class I histocompatibility antigen, B-82 alpha chain; HLA locus; HLA molecule; HLAA; HLA-A; HLA-A locus alpha 2 domain; HLA-A null; HLA-A11; HLA-A33; HLA-Aw33.1; HLAB; HLA-B; HLA-B 1502; HLA-B alpha chain; HLA-B alpha chain (B*2706); HLA | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
3106 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P01889 | |
HLA-B | |
Peptide with sequence C-DRNTQIYKAQAQT. | |
100 μg | |
Primary | |
Human | |
Liquid |