missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HLA-ABCE Monoclonal antibody specifically detects HLA-ABCE in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
Specifications
| Antigen | HLA-ABCE |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | TP25.99SF |
| Conjugate | Janelia Fluor 646 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | HLA-ABC, HLA-E, major histocompatibility complex, class I, A, major histocompatibility complex, class I, B, major histocompatibility complex, class I, C, major histocompatibility complex, class I, E, MHC class I |
| Host Species | Mouse |
| Immunogen | IFN-gamma-treated human melanoma cells COLO 38. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?