missing translation for 'onlineSavingsMsg'
Learn More

HLA-ABCE Antibody (TP25.99SF), DyLight 755, Novus Biologicals™

Product Code. 18407612 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407612 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407612 Supplier Novus Biologicals Supplier No. NBP268006IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HLA-ABCE Monoclonal antibody specifically detects HLA-ABCE in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA-ABCE
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone TP25.99SF
Conjugate DyLight 755
Formulation 50 mM Sodium Borate
Gene Alias HLA-ABC, HLA-E, major histocompatibility complex, class I, A, major histocompatibility complex, class I, B, major histocompatibility complex, class I, C, major histocompatibility complex, class I, E, MHC class I
Host Species Mouse
Immunogen IFN-gamma-treated human melanoma cells COLO 38.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.