Learn More
HLA-ABC Mouse anti-Human, APC, Clone: G46-2.6, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555555
Additional Details : Weight : 0.02000kg
Description
Reacts with all major histocompatibility Class II HLA-DR, DP and most DQ antigens expressed on B cells, antigen presenting cells and activated T cells. This antibody fixes complement. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Flow Cytometry
Specifications
HLA-ABC | |
Monoclonal | |
APC | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
G46-2.6 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.