Learn More
HLA-ABC Mouse anti-Human, APC, Clone: G46-2.6, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 562006
Additional Details : Weight : 1.00000kg
Description
The G46-2.6 monoclonal antibody specifically binds to a monomorphic epitope on the alpha chain of major histocompatibility (MHC) HLA Class I antigens. Class I MHC antigens are widely distributed on human nucleated cells.
Specifications
HLA-ABC | |
Monoclonal | |
APC | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
G46-2.6 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
For Research Use Only.