Learn More
HLA-A2 Monoclonal Antibody (BB7.2), NovaFluor™ Blue 690, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£166.00 - £354.00
Specifications
| Antigen | HLA-A2 |
|---|---|
| Clone | BB7.2 |
| Concentration | 0.6 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
This BB7.2 monoclonal antibody recognizes the human major histocompatibility complex (MHC) class I molecule HLA-A2, which is a member of the HLA-A family. MHC class I exists as a single alpha chain composed of three domains. This molecule associates with beta 2-microglobulin and is expressed on all nucleated cells. The BB7.2 monoclonal antibody has been reported to exhibit blocking activity, as well as recognize HLA-A28. Applications Tested: This BB7.2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5uL (0.6 ug) per test. A test is defined as the amount (ug) of antibody that will stain a cell sample in a final volume of 100uL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. NovaFluor dyes are not compatible with DNA intercalating viability dyes. Invitrogen LIVE/DEAD Fixable Dead Cell stains are recommended for use with NovaFluor dyes. Use 5uL of CellBlox Blocking Buffer per stained cell sample containing 10^3 to 10^8 cells. Excitation: 494 nm; Emission: 755 nm; Laser: 488 nm (Blue) Laser
HLA-A belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. Class I molecules play a central role in the immune system by presenting peptides derived from the endoplasmic reticulum lumen. They are expressed in nearly all cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domains, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exons 6 and 7 encode the cytoplasmic tail. Polymorphisms within exon 2 and exon 3 are responsible for the peptide binding specificity of each class one molecule. Typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. Hundreds of HLA-A alleles have been described. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Specifications
| HLA-A2 | |
| 0.6 μg/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| A29 protein; antigen presenting molecule; Aw-33; Aw-74; DAQB-90C11.16; FLJ26655; HLA A; HLA class I antigen; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA locus; HLAA; HLA-A; HLA-A locus alpha 2 domain; HLA-A null; HLA-A11; HLA-A33; HLA-Aw33.1; HLA-DQB1; HLA-DRB1; Human leukocyte antigen A; leucocyte antigen A; leukocyte antigen class I-A; major histocompatibility complex, class I, A; MCH class I antigen; MHC 1; MHC class 1 antigen; MHC class I anti; MHC Class I antigen; MHC class I antigen A*30; MHC class I antigen A*33; MHC class I antigen A*74; MHC class I antigen heavy chain; MHC class I antigen HLA-A heavy chain; MHC class I antigen HLA-A33; MHC class I antigen null protein; MHC class I antigene; MHC class II antigen; MHC classI antigen; MHC1; truncated MHC class I antigen | |
| HLA-A | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! |
| BB7.2 | |
| Flow Cytometry | |
| NovaFluor Blue 690 | |
| Mouse | |
| Human | |
| P04439 | |
| 3105 | |
| IgG2b κ | |
| Affinity chromatography | |
| HLA-A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.