missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA A2 Antibody (BB7.2), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-44896G
This item is not returnable.
View return policy
Description
HLA A2 Monoclonal specifically detects HLA A2 in Human samples. It is validated for Flow Cytometry.
Specifications
| HLA A2 | |
| Monoclonal | |
| DyLight 488 | |
| A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| The antibody BB7.2 recognizes an epitope at the C-terminus of alpha-2 helix and a turn on one of the underlying beta strands within the human HLA-A2 histocompatibility antigen. | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| BB7.2 | |
| Flow Cytometry | |
| HLA-A | |
| HLA-A2 solubilised by papain | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Immunology | |
| 3105 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction