Learn More
HLA-A/HLA-B/HLA-C Mouse anti-Human, Clone: SPM419, Abnova™
Mouse monoclonal antibody raised against native human HLA-A/HLA-B/HLA-C.
Brand: Abnova MAB13068.100ug
Description
HLA-C belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. Class I molecules play a central role in the immune system by presenting peptides derived from endoplasmic reticulum lumen. They are expressed in nearly all cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domain, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exons 6 and 7 encode the cytoplasmic tail. Polymorphisms within exon 2 and exon 3 are responsible for the peptide binding specificity of each class one molecule. Typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. Over one hundred HLA-C alleles have been described [provided by RefSeq]
Specifications
| HLA-A/HLA-B/HLA-C | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| HLAA | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 3105/3106/3107 | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence | |
| SPM419 | |
| Mouse monoclonal antibody raised against native human HLA-A/HLA-B/HLA-C. | |
| In 10mM PBS. | |
| HLA-A | |
| Human peripheral blood leukocytes. | |
| 100 μg | |
| Primary | |
| Reacts with a monomorphic determinant of human major histocompatibility (MHC) class I antigens (HLA-A, B and C). Human MHC class I antigens are expressed constitutively on all nucleated cells lymphocytes such as lymphocytes, thymocytes, granulocytes, and bone marrow cells and are absent on erythrocytes. MHC class I antigens play a role in class I MHC-associated antigen presentation, inhibition of NK cell cytotoxicity, tumor surveillance, and tissue allotransplantation. | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.