missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HKDC1 Recombinant Rabbit Monoclonal Antibody (K01_2M73)

Product Code. 30830061
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30830061 100 μL
30828812 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830061 Supplier Invitrogen™ Supplier No. MA564087
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

HKDC1 Recombinant Monoclonal Antibody for Western Blot

The epidermal growth factor receptor (HKDC1; ErbB-1; HER1 in humans) is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands. It is a member of the ErbB family of receptors, a subfamily of four closely related receptor tyrosine kinases: HKDC1 (ErbB-1), HER2/c-neu (ErbB-2), Her 3 (ErbB-3) and Her 4 (ErbB-4). HKDC1 exists on the cell surface and is activated by binding of its specific ligands, including epidermal growth factor and transforming growth factor alpha (TGFalpha). HKDC1 and its ligands are cell signaling molecules involved in diverse cellular functions, including cell proliferation, differentiation, motility, and survival, and in tissue development. Mutations that lead to HKDC1 overexpression (known as upregulation) or overactivity have been associated with a number of cancers, including lung cancer and glioblastoma multiforme. In this latter case a more or less specific mutation of HKDC1, called HKDC1vIII is often observed.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HKDC1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2M73
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene Hkdc1
Gene Accession No. Q2TB90
Gene Alias BC016235; hexokinase 1-like; hexokinase domain containing 1; hexokinase domain-containing protein 1; Hexokinase HKDC1; Hkdc1; LOC100364027; putative hexokinase HKDC1
Gene Symbols Hkdc1
Host Species Rabbit
Immunogen Recombinant protein of human HKDC1.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80201
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.