Learn More
Invitrogen™ HK1 Monoclonal Antibody (7A7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515675
Description
MA5-15675 targets HK1 in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Human, mouse, and Rat samples. The MA5-15675 immunogen is purified recombinant fragment of human HK1 expressed in E. Coli. MA5-15675 detects HK1 which has a predicted molecular weight of approximately 102kDa.
This gene encodes a member of the ARHGAP family of Rho/Rac/Cdc42-like GTPase activating proteins. The encoded protein interacts with the Ras-related protein Gem through its N-terminal domain. Separately, it interacts with RhoA through a RhoGAP domain, and stimulates RhoA-dependent GTPase activity. Alternatively spliced variants that encode different protein isoforms have been described but their full-length nature has not been determined.
Specifications
| HK1 | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| P05708, P17710, P19367 | |
| HK1 | |
| Purified recombinant fragment of human HK1 expressed in E. Coli. | |
| RUO | |
| 15275, 25058, 3098 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 7A7 | |
| Unconjugated | |
| HK1 | |
| BB404130; brain form hexokinase; dea; downeast anemia; glycolytic enzyme; HEXOKIN; hexokinase; hexokinase 1; hexokinase I; hexokinase IR; hexokinase type I; Hexokinase, tumor isozyme; hexokinase-1; Hexokinase-A; HK; HK I; HK1; Hk-1; Hk1-s; HK1-ta; HK1-tb; HK1-tc; HKD; HKI; HMSNR; HXK1; mHk1-s | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.