missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HJURP Recombinant Rabbit Monoclonal Antibody (K01_2M72)

Product Code. 30828564
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828564 100 μL
30828854 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828564 Supplier Invitrogen™ Supplier No. MA564086

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

HJURP Recombinant Monoclonal Antibody for IHC (P), Western Blot

Holliday junction recognition protein (HJURP) is a centromeric protein that plays a central role in the incorporation and maintenance of histone H3-like variant CENPA at centromeres. It acts as a specific chaperone for CENPA and is required for the incorporation of newly synthesized CENPA molecules into nucleosomes at replicated centromeres. HJURP prevents CENPA-H4 tetramerization and prevents premature DNA binding by the CENPA-H4 tetramer. It directly binds to Holliday junctions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HJURP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_2M72
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene HJURP
Gene Accession No. Q8NCD3
Gene Alias 14-3-3-associated AKT substrate; 6430706D22Rik; A730008H23Rik; C330011F01Rik; FAKTS; fetal liver expressing gene 1; fetal liver expressing gene 1 protein homolog; Fetal liver-expressing gene 1 protein; FLEG1; Gm975; hFLEG1; Hjurp; Holliday junction recognition protein; mFleg1; RGD1305846; Up-regulated in lung cancer 9; URLC9
Gene Symbols HJURP
Host Species Rabbit
Immunogen Recombinant protein of human HJURP.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55355
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.