Learn More
Invitrogen™ HJURP Recombinant Rabbit Monoclonal Antibody (16H6L17)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 703460
Description
This antibody is predicted to react with Monkey, Pig, Cat. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Holliday junction recognition protein (HJURP) is a centromeric protein that plays a central role in the incorporation and maintenance of histone H3-like variant CENPA at centromeres. It acts as a specific chaperone for CENPA and is required for the incorporation of newly synthesized CENPA molecules into nucleosomes at replicated centromeres. HJURP prevents CENPA-H4 tetramerization and prevents premature DNA binding by the CENPA-H4 tetramer. It directly binds to Holliday junctions.
Specifications
| HJURP | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8NCD3 | |
| HJURP | |
| Proteins corresponding to human HJURP [1) aa47-aa177 2)aa394-aa748]. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry, Western Blot | |
| 16H6L17 | |
| Unconjugated | |
| HJURP | |
| 14-3-3-associated AKT substrate; 6430706D22Rik; A730008H23Rik; C330011F01Rik; FAKTS; fetal liver expressing gene 1; fetal liver expressing gene 1 protein homolog; Fetal liver-expressing gene 1 protein; FLEG1; Gm975; hFLEG1; Hjurp; Holliday junction recognition protein; mFleg1; RGD1305846; Up-regulated in lung cancer 9; URLC9 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 55355 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.