missing translation for 'onlineSavingsMsg'
Learn More

HJURP Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA552562

Additional Details : Weight : 0.02000kg

Product Code. 15760511

  • £427.00 / 100µL
Estimated Shipment: 21-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: TPVVQMATLT YETPQGLRIW GGRLIKERNE GEIQDSSMKP ADRTDGSVQA AAWGPELPSH RTVLGADSKS GEVDATSDQE ESVAWALAPA VPQSPLKNEL RRKYLTQVDI LLQGAEYFEC AGNRAGRDVR V Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 43%.

HJURP Polyclonal antibody specifically detects HJURP in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

HJURP
Polyclonal
Unconjugated
HJURP
14-3-3-associated AKT substrate; 6430706D22Rik; A730008H23Rik; C330011F01Rik; FAKTS; fetal liver expressing gene 1; fetal liver expressing gene 1 protein homolog; Fetal liver-expressing gene 1 protein; FLEG1; Gm975; hFLEG1; Hjurp; Holliday junction recognition protein; mFleg1; RGD1305846; Up-regulated in lung cancer 9; URLC9
Rabbit
Antigen affinity chromatography
RUO
55355
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8NCD3
HJURP
Recombinant protein corresponding to Human HJURP
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only