missing translation for 'onlineSavingsMsg'
Learn More

HJURP Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18413737 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413737 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413737 Supplier Novus Biologicals Supplier No. NBP147288AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HJURP Polyclonal antibody specifically detects HJURP in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HJURP
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias FAKTS14-3-3-associated AKT substrate, fetal liver expressing gene 1, Fetal liver-expressing gene 1 protein, FLEG1, hFLEG1, Holliday junction recognition protein, Up-regulated in lung cancer 9, URLC9DKFZp762E1312
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 700 and 748 of human Holliday Junction Recognition Protein using the numbering given in entry NP_060880.3 (GeneID 55355).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55355
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.