missing translation for 'onlineSavingsMsg'
Learn More

HJB97, MedChemExpress

Product Code. 16440927
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16440927

Brand: Medchem Express HY112429

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HJB97 is a high-affinity BET inhibitor with Kis of 0.9 nM (BRD2 BD1), 0.27 nM (BRD2 BD2), 0.18 nM (BRD3 BD1), 0.21 nM (BRD3 BD2), 0.5 nM (BRD4 BD1), 1.0 nM (BRD4 BD2), respectively. HJB97 is employed for the design of potential PROTAC BET degrader and has antitumor activity.

HJB97 is a high-affinity BET inhibitor with Kis of 0.9 nM (BRD2 BD1), 0.27 nM (BRD2 BD2), 0.18 nM (BRD3 BD1), 0.21 nM (BRD3 BD2), 0.5 nM (BRD4 BD1), 1.0 nM (BRD4 BD2), respectively. HJB97 is employed for the design of potential PROTAC BET degrader and has antitumor activity.

Specifications

Chemical Name or Material HJB97
CAS 2093391-24-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C26H28N8O3
Quantity 50 mg
Solubility Information DMSO : 30 mg/mL (59.93 mM; Need ultrasonic)
SMILES O=C(C1=NC(NC2=CC(C3CC3)=NN2CC)=C4C(NC5=C4C=C(OC)C(C6=C(C)ON=C6C)=C5)=N1)NC
Molecular Weight (g/mol) 500.55
Formula Weight 500.55
Percent Purity 98.24%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.