missing translation for 'onlineSavingsMsg'
Learn More

HJ 921-2017 Organochlorine Pesticides Mixture 109 100 μg/mL in Toluene:n-Hexane, Dr. Ehrenstorfer

Product Code. 30315364
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315364 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315364 Supplier Dr. Ehrenstorfer Supplier No. DREA50000109TH

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4'-DDD∣2,4'-DDE∣2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Cis-Chlordane (Alpha Isomer)∣cis-Nonachlor∣delta-HCH∣Dieldrin∣Endrin∣gamma-HCH∣Heptachlor-endo-epoxide (trans-, isomer A)∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hexachlorobenzene∣Mirex∣Trans-Chlordane (Gamma Isomer)∣trans-Nonachlor
CAS 53-19-0, 3424-82-6, 789-02-6, 72-54-8, 72-55-9, 50-29-3
Analyte or Component Names 2,4'-DDD∣2,4'-DDE∣2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Cis-Chlordane (Alpha Isomer)∣cis-Nonachlor∣delta-HCH∣Dieldrin∣Endrin∣gamma-HCH∣Heptachlor-endo-epoxide (trans-, isomer A)∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hexachlorobenzene∣Mirex∣Trans-Chlordane (Gamma Isomer)∣trans-Nonachlor
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix n-Hexane/Toluene
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2,4'-DDD 100 μg/mL∣2,4'-DDE 100 μg/mL∣2,4'-DDT 100 μg/mL∣4,4'-DDD 100 μg/mL∣4,4'-DDE 100 μg/mL∣4,4'-DDT 100 μg/mL∣alpha-Endosulfan 100 μg/mL∣alpha-HCH 100 μg/mL∣beta-Endosulfan 100 μg/mL∣beta-HCH 100 μg/mL∣Cis-Chlordane (Alpha Isomer) 100 μg/mL∣cis-Nonachlor 100 μg/mL∣delta-HCH 100 μg/mL∣Dieldrin 100 μg/mL∣Endrin 100 μg/mL∣gamma-HCH 100 μg/mL∣Heptachlor-endo-epoxide (trans-, isomer A) 100 μg/mL∣Heptachlor-exo-epoxide (cis-, isomer B) 100 μg/mL∣Hexachlorobenzene 100 μg/mL∣Mirex 100 μg/mL∣Trans-Chlordane (Gamma Isomer) 100 μg/mL∣trans-Nonachlor 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.