missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HJ 867-2017, YC/T333-2010 Phthalate Esters Mixture 157 5000 μg/mL in n-Hexane, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| CAS | 84-74-2, 84-66-2, 117-84-0, 85-68-7, 117-81-7, 84-69-5 |
| Quantity | 1 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | n-Hexane |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction