missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HJ 834-2017, HJ 951-2018 VOC & SVOCs Internal Standards Mixture 174 1000 μg/mL in Acetone/Dichloromethane, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | 1,4-Dichlorobenzene-D4∣Acenaphthene D10∣Chrysene D12∣Naphthalene D8∣Perylene D12∣Phenanthrene D10 |
| CAS | 3855-82-1, 15067-26-2, 1719-03-5, 1146-65-2, 1520-96-3, 1517-22-2 |
| Solvent or Matrix | Acetone/Dichloromethane |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | 1,4-Dichlorobenzene-D4 1000 μg/ mL∣Acenaphthene D10 1000 μg/ mL∣Chrysene D12 1000 μg/ mL∣Naphthalene D8 1000 μg/ mL∣Perylene D12 1000 μg/ mL∣Phenanthrene D10 1000 μg/ mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction