missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HJ 827-2017 Carbamate Pesticides Mixture 520 50-1000 μg/mL in Methanol, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | 2,3,5-Trimethacarb∣Bendiocarb∣Carbanolate (Banol)∣Carbaryl∣Carbofuran∣Carbofuran-3-hydroxy∣Fenobucarb∣Isoprocarb∣Methiocarb∣Methomyl∣Methomyl-oxime∣Metolcarb∣Pirimicarb∣Promecarb∣Propoxur |
| CAS | 2655-15-4, 22781-23-3, 671-04-5, 63-25-2, 1563-66-2, 16655-82-6 |
| Analyte or Component Names | 2,3,5-Trimethacarb∣Bendiocarb∣Carbanolate (Banol)∣Carbaryl∣Carbofuran∣Carbofuran-3-hydroxy∣Fenobucarb∣Isoprocarb∣Methiocarb∣Methomyl∣Methomyl-oxime∣Metolcarb∣Pirimicarb∣Promecarb∣Propoxur |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Carbofuran 50 μg/mL∣Isoprocarb 50 μg/mL∣Methiocarb 50 μg/mL∣Pirimicarb 50 μg/mL∣Carbanolate (Banol) 100 μg/mL∣Fenobucarb 100 μg/mL∣Metolcarb 100 μg/mL∣Promecarb 100 μg/mL∣2,3,5-Trimethacarb 200 μg/mL∣Bendiocarb 200 μg/mL∣Carbaryl 200 μg/mL∣Carbofuran-3-hydroxy 200 μg/mL∣Methomyl 200 μg/mL∣Propoxur 200 μg/mL∣Methomyl-oxime 1000 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction