missing translation for 'onlineSavingsMsg'
Learn More

HJ 827-2017 Carbamate Pesticides Internal Standards Mixture 177 25-100 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30320872
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30320872 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30320872 Supplier Dr. Ehrenstorfer Supplier No. DREA50000177AC
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbaryl-d7∣Carbofuran-d3∣Methiocarb D3 (N-methyl D3)∣Methomyl D3
CAS 362049-56-7, 1007459-98-4, 1581694-94-1, 1398109-07-3
Analyte or Component Names Carbaryl-d7∣Carbofuran-d3∣Methiocarb D3 (N-methyl D3)∣Methomyl D3
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetone
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Methiocarb D3 (N-methyl D3) 25 μg/mL∣Carbaryl-d7 100 μg/mL∣Carbofuran-d3 100 μg/mL∣Methomyl D3 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.