missing translation for 'onlineSavingsMsg'
Learn More

HJ 699-2014 Organochlorine Pesticides Mixture 517 10 μg/mL in n-Hexane, Dr. Ehrenstorfer

Product Code. 30324856
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324856 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324856 Supplier Dr. Ehrenstorfer Supplier No. DREA50000517HE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4'-DDD∣2,4'-DDE∣2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Cis-Chlordane (Alpha Isomer)∣delta-HCH∣Dicofol∣Dieldrin∣Endosulfan-sulfate∣Endrin∣Endrin-aldehyde∣Endrin-ketone∣gamma-HCH∣Heptachlor∣Heptachlor-endo-epoxide (trans-, isomer A)∣Heptachlor-exo-epoxide (cis-, isomer B)∣Methoxychlor∣Quintozene∣Trans-Chlordane (Gamma Isomer)
CAS 53-19-0, 3424-82-6, 789-02-6, 72-54-8, 72-55-9, 50-29-3
Analyte or Component Names 2,4'-DDD∣2,4'-DDE∣2,4'-DDT∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Cis-Chlordane (Alpha Isomer)∣delta-HCH∣Dicofol∣Dieldrin∣Endosulfan-sulfate∣Endrin∣Endrin-aldehyde∣Endrin-ketone∣gamma-HCH∣Heptachlor∣Heptachlor-endo-epoxide (trans-, isomer A)∣Heptachlor-exo-epoxide (cis-, isomer B)∣Methoxychlor∣Quintozene∣Trans-Chlordane (Gamma Isomer)
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix n-Hexane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2,4'-DDD 10 μg/mL∣2,4'-DDE 10 μg/mL∣2,4'-DDT 10 μg/mL∣4,4'-DDD 10 μg/mL∣4,4'-DDE 10 μg/mL∣4,4'-DDT 10 μg/mL∣Aldrin 10 μg/mL∣alpha-Endosulfan 10 μg/mL∣alpha-HCH 10 μg/mL∣beta-Endosulfan 10 μg/mL∣beta-HCH 10 μg/mL∣Cis-Chlordane (Alpha Isomer) 10 μg/mL∣delta-HCH 10 μg/mL∣Dicofol 10 μg/mL∣Dieldrin 10 μg/mL∣Endosulfan-sulfate 10 μg/mL∣Endrin 10 μg/mL∣Endrin-aldehyde 10 μg/mL∣Endrin-ketone 10 μg/mL∣gamma-HCH 10 μg/mL∣Heptachlor 10 μg/mL∣Heptachlor-endo-epoxide (trans-, isomer A) 10 μg/mL∣Heptachlor-exo-epoxide (cis-, isomer B) 10 μg/mL∣Methoxychlor 10 μg/mL∣Quintozene 10 μg/mL∣Trans-Chlordane (Gamma Isomer) 10 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.