missing translation for 'onlineSavingsMsg'
Learn More

HJ 642-2013 VOC Internal Standards Mixture 134 250 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30326057
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326057 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30326057 Supplier Dr. Ehrenstorfer Supplier No. DREA50000134ME
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Bromofluorobenzene∣Toluene D8
CAS 460-00-4, 2037-26-5
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 4-Bromofluorobenzene 250 μg/mL∣Toluene D8 250 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.