missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HJ 350-2007 PCB Mixture 680 10 μg/mL in Isooctane, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| CAS | 2437-79-8, 16605-91-7, 2051-60-7, 37680-73-2, 38380-03-9, 35065-28-2 |
| Quantity | 1 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Isooctane |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction