missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIVEP2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-46125
This item is not returnable.
View return policy
Description
HIVEP2 Polyclonal specifically detects HIVEP2 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Peptide ELISA.
Specifications
| HIVEP2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| c-myc intron binding protein 1, c-myc intron-binding protein 1, human immunodeficiency virus type I enhancer binding protein 2, Human immunodeficiency virus type I enhancer-binding protein 2HIV-EP2MBP-2SHN2, MHC binding protein-2, MHC-binding protein 2, MIBP1, Schnurri-2, transcription factor HIVEP2, ZAS2, ZNF40B | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3097 | |
| Human, Mouse, Rat, Bovine, Canine | |
| IgG |
| ELISA | |
| 0.5 mg/mL | |
| Peptide ELISA Detection limit 1:1000 | |
| NP_006725.3 | |
| HIVEP2 | |
| Peptide with sequence C-TEESKDPSSEKSQ corresponding to C-Terminus according to NP_006725.3. | |
| 0.1 mg | |
| Primary | |
| This antibody is specific to the C-Terminus. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction