missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIV1 p24 Monoclonal Antibody (2N32)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA171521
This item is not returnable.
View return policy
Description
This antibody reacts with HIV-1 p24 subtype B, C and A. It has been mapped to amino acids 80-90 (WDRVHPVHAGP - B subtype consensus). This clone may also be referred to as 2N32 (05-009).
HIV-1 Gag p24 is a capsid protein that constitutes the core of the AIDS virus HIV-1 and is produced by the digestion of its precursor Gag p55 by HIV-1 protease. The HIV-1 Gag p24 protein is indispensable to the reproduction of AIDS virus, and constitutes an essential element for the AIDS virus particle construction. HIV-1 Gag p24 is detectable from the early stage of AIDS virus infection, and is used as a marker for observation of the development in the patient's condition after treatment, as it indicated the amount of virus in the blood.
Specifications
| HIV1 p24 | |
| Monoclonal | |
| 0.6 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| 2N32 | |
| Unconjugated | |
| HIV p24; HIV1gp1 | |
| Recombinant p24 protein of HIV-1 B-subtype consensus purified from E. coli. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction