missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIV1 p24 (group O, strain BCF06) Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581808
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for HIV-1 p24 Protein (group O, strain BCF06).
HIV-1 Gag p24 is a capsid protein that constitutes the core of the AIDS virus HIV-1 and is produced by the digestion of its precursor Gag p55 by HIV-1 protease. The HIV-1 Gag p24 protein is indispensable to the reproduction of AIDS virus, and constitutes an essential element for the AIDS virus particle construction. HIV-1 Gag p24 is detectable from the early stage of AIDS virus infection, and is used as a marker for observation of the development in the patient's condition after treatment, as it indicated the amount of virus in the blood.
Specifications
| HIV1 p24 (group O, strain BCF06) | |
| Polyclonal | |
| Unconjugated | |
| HIV p24; HIV1gp1 | |
| Recombinant HIV-1 p24 Protein (group O, strain BCF06) protein. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction