missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIV1 Integrase Monoclonal Antibody (IN-2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA518031
This item is not returnable.
View return policy
Description
This target has a predicted molecular weight of 32 kDa.
Specifications
| HIV1 Integrase | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Mouse | |
| Protein G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 κ |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| IN-2 | |
| Unconjugated | |
| HIV-1 | |
| Bacterially expressed, hexahistidine amino-terminal tagged HIV-1 integrase (IN) protein (clade B, HXB-3 isolate). | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction