missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences HIV pol peptide ILKEPVHGV (HLA-A*0201)
Single peptide synthesized as presented by the MHC class I allele
Brand: IBA Lifesciences 6-7005-901
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Specifications
Recommended: -25°C to -15°C | |
1 mg |