missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIV-1 Tat specific factor 1 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-06041
This item is not returnable.
View return policy
Description
HIV-1 Tat specific factor 1 Polyclonal antibody specifically detects HIV-1 Tat specific factor 1 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| HIV-1 Tat specific factor 1 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| cofactor required for Tat activation of HIV-1 transcription, dJ196E23.2, HIV TAT specific factor 1, HIV Tat-specific factor 1, HIV-1 Tat specific factor 1, TAT-SF1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 27336 | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_055315.2 | |
| HTATSF1 | |
| Peptide with sequence C-QELYGDGKDGDTQTD corresponding to internal region of HIV-1 Tat specific factor 1 (near N-Terminus) according to NP_055315.2. | |
| 0.1 mg | |
| Primary | |
| Cross reacts with Human. Expected from sequence similarity: Rat and Dog. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction