missing translation for 'onlineSavingsMsg'
Learn More

HIV-1 Tat specific factor 1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18413404 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413404 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413404 Supplier Novus Biologicals Supplier No. NBP118912JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HIV-1 Tat specific factor 1 Polyclonal antibody specifically detects HIV-1 Tat specific factor 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIV-1 Tat specific factor 1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias cofactor required for Tat activation of HIV-1 transcription, dJ196E23.2, HIV TAT specific factor 1, HIV Tat-specific factor 1, HIV-1 Tat specific factor 1, TAT-SF1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 375 and 425 of human HIV-1 Tat specific factor 1 using the numbering given in entry NP_055315.2 (GeneID 27336).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 27336
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.