missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIV-1 Rev binding protein (HRB) Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Shop All Bio Techne ProductsDescription
HIV-1 Rev binding protein (HRB) Polyclonal specifically detects HIV-1 Rev binding protein (HRB) in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | HIV-1 Rev binding protein (HRB) |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | ArfGAP with FG repeats 1, DKFZp686I15205, HIV-1 Rev-binding protein, HRBRev interacting protein, Rab, Rev/Rex activation domain-binding protein, RABMGC116938, RIPMGC116940 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human HIV-1 Rev binding protein (HRB) (NP_004495.2). |
| Purification Method | Affinity purified |
| Quantity | 100 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?