missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIV-1 Protease Antibody (1696) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-337
This item is not returnable.
View return policy
Description
HIV-1 Protease Monoclonal antibody specifically detects HIV-1 Protease in Virus samples. It is validated for Western Blot, ELISA, Dot Blot.
Specifications
| HIV-1 Protease | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 0.5 ug/ml, ELISA 1:2000, Functional, Dot Blot 1:100-1:2000 | |
| Bacterially expressed full-length HIV-1 protease | |
| 0.1 mg | |
| Virology Bacteria and Parasites | |
| The antibody 1696 recognizes free N-terminus of mature HIV protease (HIV-1 and HIV-2), an enzyme that hydrolyzes polyproteins of HIV viruses into functional proteins. The antibody 1696 does not react with the precursor. | |
| Store at 4C. Do not freeze. |
| Western Blot, ELISA, Functional Assay, Dot Blot | |
| 1696 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Virus | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction