missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIV-1 Protease Antibody (1696), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-337AF700
This item is not returnable.
View return policy
Description
HIV-1 Protease Monoclonal antibody specifically detects HIV-1 Protease in Virus samples. It is validated for Western Blot, ELISA.
Specifications
| HIV-1 Protease | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Bacterially expressed full-length HIV-1 protease | |
| 0.1 mL | |
| Primary | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| 1696 | |
| Western Blot, ELISA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| The antibody 1696 recognizes free N-terminus of mature HIV protease (HIV-1 and HIV-2), an enzyme that hydrolyzes polyproteins of HIV viruses into functional proteins. The antibody 1696 does not react with the precursor. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction