missing translation for 'onlineSavingsMsg'
Learn More

HIV-1 gp120 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18433596 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433596 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433596 Supplier Novus Biologicals Supplier No. NBP281822UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HIV-1 gp120 Polyclonal antibody specifically detects HIV-1 gp120 in Virus samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIV-1 gp120
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias Envelope surface glycoprotein gp120, gp120, gp120 glycoprotein, Human immunodeficiency virus 1, SU, Surface protein
Host Species Rabbit
Immunogen HIV-1 gp120 antibody was raised against a peptide corresponding to 15 amino acids near the center of HIV-1 gp120. Amino Acid Squence: VAPTKAKRRVVQREKR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 155971
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.