missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HIV-1 gp120 Polyclonal antibody specifically detects HIV-1 gp120 in Virus samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | HIV-1 gp120 |
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 647 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Envelope surface glycoprotein gp120, gp120, gp120 glycoprotein, Human immunodeficiency virus 1, SU, Surface protein |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant HIV-1 gp120 (Ala 29-Glu 503; 97CN001 strain). |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Haben Sie Verbesserungsvorschläge?