missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ HIV-1 Gag p24 Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Description
The HIV-1 Gag p24 Quantikine ELISA Kit quantifies viral HIV-1 Gag p24 in cell culture supernates (100 μL).
Specifications
Specifications
| Assay Range | 7.81 to 500 pg/mL |
| Assay Sensitivity | 0.24 pg/mL |
| Sample Type | Cell Culture Supernatant |
| Gene ID (Entrez) | 155030 |
| Host Species | Viral |
| Quantity | 1 Kit |
| Sample Volume | Cell Culture Supernates (100 μL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Total Assay Time | 4.5 hr. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction