missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Protein G HP Prepacked Columns
Use for fast, easy separations of IgG from different species and subclasses. Cytiva Life Sciences™ HiTrap™ Protein G HP Prepacked Columns are prepacked with Protein G Sepharose™.
Brand: Cytiva 17-0404-03
Code : GE
Additional Details : Weight : 0.13800kg
Description
- Prepacked with Protein G Sepharose High Performance
- Binding capacity: 25mg human IgG/column
- Maximum pressure: 3 bar
- Column material: polypropylene
- For use with syringe, peristaltic pump, or chromatography system
- 1 and 5mL columns
Media
- Ligand: Recombinant protein G lacking albumin-binding region
- Matrix: 6% cross-linked agarose
- Average particle size: 34μm
- pH stability working range: 3 to 9
1mL columns
- Bed dimensions: 7 x 25mm
- Flow velocity: <4mL/min.
5mL columns
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
Specifications
Pre-packed Columns | |
7 mm | |
>4 mL/min. | |
Designed for use with a syringe, peristaltic pump, or chromatography system | |
20% Ethanol | |
2 Pk. |
25 mg Human IgG/Column | |
0.27 in. | |
1 mL | |
HiTrap Protein G HP are prepacked columns for general-purpose preparative purification of monoclonal and polyclonal antibodies from most species including rat. | |
4°C to 30°C |