Learn More
Cytiva HiTrap MabSelect PrismA Column
Protein A Column
Brand: Cytiva 17549854
Additional Details : CAS Number : 64-17-5 Weight : 0.20000kg
Description
These columns are prepacked with MabSelect PrismA protein A chromatography resin that has been improved with an optimized high-flow agarose base matrix and a genetically engineered protein A-derived ligand, allowing efficient cleaning between purification runs.
Specifications
5 mL | |
5 bar (0.5 MPa, 70 psi) | |
Antibody affinity chromatography |
<20 mL/min. | |
5 x 5 mL | |
Pre-packed Column |
Safety and Handling
- HiTrap MabSelect PrismA
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE- per cent of the mixture consists of component(s) of unknown acute toxicity.