missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap KappaSelect Media

Product Code. 11380212 Shop All Cytiva Products
Change view
Click to view available options
Bed Volume (Metric):
1 mL
5 mL
Unit Size:
Each
Pack of 5
Product Code. Bed Volume (Metric) unitSize
11380212 5 mL Each
14811611 1 mL Pack of 5
2 options
This item is not returnable. View return policy

Product Code. 11380212

Brand: Cytiva 17545812

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Quantity 5 x 1 mL
Stability Stable in all commonly used aqueous buffers
Storage Requirements 4 to 8 deg.C, 20% Ethanol
Binding Capacity 15 mg Fab/mL of medium2)
Product Type HiTrap KappaSelect Media
Particle Size 75 μm
Material Polypropylene
Max. Pressure 300 kPa
Matrix Highly Cross-linked Agarose
pH Range 3 to 10
Bed Volume (Metric) 5 mL
Ligand Type Recombinant protein (Mr 13 000)
Show More Show Less
compliance-icons
Product Identifier
  • HiTrap 1 x 5 ml, KappaSelect
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.