missing translation for 'onlineSavingsMsg'
Learn More

Cytiva™ HiTrap™ Heparin HP Columns

Product Code. 10298944 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 5mL
5 x 1mL
5 x 5mL
Flow Rate:
<20 mL/min.
<4 mL/min.
Diameter (Metric) Inner:
0.7cm
1.6cm
Unit Size:
Each
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Flow Rate Diameter (Metric) Inner Unit Size
10298944 1 x 5mL <20 mL/min. 1.6cm Each
11738448 5 x 5mL <20 mL/min. 1.6cm Pack of 5
10288944 5 x 1mL <4 mL/min. 0.7cm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10298944 Supplier Cytiva™ Supplier No. 17040701
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Purify DNA-binding proteins, coagulation factors, lipoproteins and protein synthesis factors. Cytiva Life Sciences™ HiTrap™ Heparin HP Columns are prepacked with Heparin Sepharose™ High Performance for high-resolution purification.

  • Stable over pH range 5 to 10
  • Tolerates commonly used aqueous buffers
  • Heparin Sepharose High Performance medium links porcine heparin ligand to 6% agarose by reductive amination

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 x 5mL
Flow Rate <20 mL/min.
Particle Size 34 μm
For Use With (Equipment) Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system
Bed Volume 5mL
Product Type Heparin HP Column
Diameter (Metric) Inner 1.6cm
For Use With (Application) Purification of Antithrombin III, coagulation facto rs and other Plasma Proteins
Height (English) 0.98 in.
Height (Metric) 2.5cm
Ligand Type 10mg Heparin/mL medium
Material Polypropylene
Max. Pressure 42psi
pH Range 5 to 10
Support Type 6% Highly Cross-linked Agarose
Sample Type DNA-binding Proteins, Coagulation Factors, Lipoproteins, Protein Synthesis Factors
Content And Storage 4° to 30°C
Show More Show Less
compliance-icons
Product Identifier
  • HiTrap Heparin HP 5 ml, 1 x 5 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.