missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ GCSFSelect
Prepacked columns specifically designed for the purification of recombinant Granulocyte-colony stimulating factor (G-CSF). Based on a highly rigid agarose base matrix that allows for high flow rates at large production scales.
£784.00 - £822.00
Specifications
For Use With | Purification of Recombinant Granulocyte-Colony Stimulating Factor (G-CSF) |
---|---|
Ligand Type | Single-Chain Antibody Fragment Directed Against G-CSF |
Particle Size | 75 μm |
pH | 2 to 12 |
Quantity | 5/Pk. |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
15539271
|
Cytiva
17548311 |
1 mL |
£822.00
Pack of 5 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15549271
|
Cytiva
17548312 |
5 mL |
£784.00
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
Description
- Granulocyte-colony stimulating factor (G-CSF) is a hormone that stimulates production of white blood cells in bone marrow
- For a highly selective purification step, the affinity ligand is based on a single-chain antibody fragment directed against G-CSF
- To facilitate binding of the target molecule, the ligand is attached to the base matrix through a hydrophilic spacer arm
- Ligand is produced in a yeast expression system where fermentation, subsequent purification, and formulation are performed in the absence of animal-derived components
- GCSFSelect's rigid agarose base matrix allows for processing of large sample volumes
- Composition: Ligand coupled to Capto via stable amide bonds
Specifications
Purification of Recombinant Granulocyte-Colony Stimulating Factor (G-CSF) | |
75 μm | |
5/Pk. | |
Granulocyte-Colony Stimulating Factor (G-CSF) |
Single-Chain Antibody Fragment Directed Against G-CSF | |
2 to 12 | |
Column |