missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ GCSFSelect

Product Code. 15549271 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
1 mL
5 mL
Unit Size:
Each
Pack of 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15549271 5 mL Each
15539271 1 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15549271 Supplier Cytiva Supplier No. 17548312
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prepacked columns specifically designed for the purification of recombinant Granulocyte-colony stimulating factor (G-CSF). Based on a highly rigid agarose base matrix that allows for high flow rates at large production scales.

  • Granulocyte-colony stimulating factor (G-CSF) is a hormone that stimulates production of white blood cells in bone marrow
  • For a highly selective purification step, the affinity ligand is based on a single-chain antibody fragment directed against G-CSF
  • To facilitate binding of the target molecule, the ligand is attached to the base matrix through a hydrophilic spacer arm
  • Ligand is produced in a yeast expression system where fermentation, subsequent purification, and formulation are performed in the absence of animal-derived components
  • GCSFSelect's rigid agarose base matrix allows for processing of large sample volumes
  • Composition: Ligand coupled to Capto via stable amide bonds
TRUSTED_SUSTAINABILITY

Specifications

Flow Rate 20 mL/min
For Use With Purification of Recombinant Granulocyte-Colony Stimulating Factor (G-CSF)
Ligand Type Single-Chain Antibody Fragment Directed Against G-CSF
Particle Size 75 μm
pH 2 to 12
Quantity 5/Pk.
Binding Capacity 19.5 mg/column
Volume (Metric) 5 mL
Purification Target Granulocyte-Colony Stimulating Factor (G-CSF)
Product Type Column
compliance-icons
Product Identifier
  • HiTrap GCSFSelect 5 ml, 1 x 5 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.