missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap Chelating Sepharose FF Media

Product Code. 13485177 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13485177 5 x 1 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13485177 Supplier Cytiva Supplier No. 90100347
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Quantity 5 x 1 mL
Stability Stable in all commonly used aqueous buffers
Storage Requirements 4 to 30 deg.C, 20% Ethanol
Product Type HiTrap Chelating Sepharose FF Media
Particle Size 45 to 165 μm
Material Polypropylene
Max. Pressure 100 kPa
Matrix 6% Cross-linked Agarose
pH Range 3 to 13
Bed Volume (Metric) 1 mL
Ligand Type Iminodiacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • HiTrap Chelating Sepharose 6 Fast Flow, 5 x 1 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.