missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Capto™ S Column

Product Code. 16380896 Shop All Cytiva Products
Change view
Click to view available options
Length (Metric):
25 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
16380896 25 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16380896 Supplier Cytiva Supplier No. 29400458
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Prepacked with a strong cation exchange modern resin for small scale protein purification

  • Capto S resin couples a sulfoethyl strong cation exchanger to a chemically modified, high-flow agarose matrix
  • The rigid agarose matrix provides greater particle rigidity without compromising pore size, which allows for high bed volume and purification of viscous samples at high flow rates
  • High dynamic binding capacity is achieved with a dextran surface extender that coats the agarose matrix
  • Combining high binding capacity, high flow rate, and low back pressure reduces process cycle times and improves productivity at process scale
  • Suitable for screening of selectivity, binding and elution conditions, and small-scale purifications
  • Optimized for capture and intermediate protein purification
  • Convenient HiTrap format for easy connection to a syringe, peristaltic pump, or chromatography system such as an ÄKTA system
TRUSTED_SUSTAINABILITY

Specifications

Column Format HiTrap™ Column
Diameter (Metric) 7 mm
Flow Rate <4 mL/min
For Use With Small Scale Protein Purification
Ion Exchange Group Strong Cation
Length (Metric) 25 mm
Ligand Type Sulfoethyl (S)
Max. Pressure 5 bar (70 psi)
pH 4 to 12
Quantity 1 Ea.
Matrix Capto
Volume (Metric) 1 mL
Technique Ion Exchange Chromatography
Product Type Prepacked Column
Show More Show Less
compliance-icons
Product Identifier
  • HiTrap Capto S, 1 x 1 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-18 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.