missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Capto™ MMC Column

Product Code. 16340906 Shop All Cytiva Products
Click to view available options
Length (Metric):
25 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16340906

Brand: Cytiva 29400465

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Prepacked with a multimodal ion exchange chromatography resin for direct load of clarified feedstocks

  • Prepacked with a multimodal BioProcess ion exchange resin that offers a different selectivity compared to traditional ion exchangers
  • Binds proteins at high or low ionic strength
  • Resin may be used for direct load of clarified feedstocks, without prior dilution to reduce the conductivity of the starting material
  • Can be used to bind proteins at the conductivity of the feed material, and to solve specific purification challenges
  • To minimize tedious sample preparation, the top filter in HiTrap Capto MMC is optimized for loading sonicated unclarified cell lysate directly on the column
  • The ligand contains a carboxyl group, making it a weak cation exchanger, which may cause it to interact with proteins in many different ways
  • High dynamic binding capacity at high conductivity
  • Selectivity allows binding in the presence of salt
  • High volume throughput
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 7 mm
Flow Rate <4 mL/min
For Use With Direct Load of Clarified Feedstocks
Ion Exchange Group Multimodal Weak Cation
Length (Metric) 25 mm
Max. Pressure 5 bar (70 psi)
Particle Size 75 μm
pH 2 to 14
Quantity 1 Ea.
Temperature 4°C to 30°C
Content And Storage Store at 4°C to 30°C in 20% Ethanol
Binding Capacity >45 mg BSA/mL medium
Matrix Highly Cross-Linked Agarose
Volume (Metric) 1 mL
Type Prepacked Column
Technique Multimodal Ion Exchange Chromatography
Show More Show Less
compliance-icons
Product Identifier
  • HiTrap Capto MMC, 1 x 1 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.