Learn More
Cytiva HiTrap™ Capto™ Core 700 Column
Prepacked with multimodal resin for intermediate purification, polishing of viruses, and large molecules
Brand: Cytiva 29400461
Additional Details : Weight : 0.05000kg
Description
- Prepacked HiTrap columns for easy screening and convenient process development, as well as small scale purification
- Capto Core 700 is a novel, layered bead chromatography resin consisting of a porous outer layer and ligand-activated core
- The octylamine ligands found in the core of the beads are multimodal, being both hydrophobic and positively charged
- Internalized ligands bind impurities strongly over a wide range of pH and salt concentrations
- The pores of the bead outer layer prevent large targets (Mr > 700,000) from binding to the ligands while smaller impurities can enter freely into the beads where they are captured
- High capacity and productivity in flowthrough mode
- Wide operational window of pH and conductivity
Specifications
HiTrap™ Column | |
<4 mL/min | |
5 bar (70 psi) | |
1 Ea. | |
Store at 4°C to 30°C, 20% Ethanol | |
High Flow Agarose | |
Prepacked Column |
7 mm | |
25 mm | |
2 to 14 | |
4°C to 30°C | |
∼13 mg ovalbumin/mL medium | |
1 mL | |
Multimodal Ion Exchange Chromatography |
Safety and Handling
- HiTrap Capto Core 700, 1 x 1 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.