missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Capto™ Adhere Column
Shop All Cytiva Products
Click to view available options
Length (Metric):
25 mm
Unit Size:
Each
Description
- A strong anion exchanger with multimodal functionality
- Designed for post-protein A purification of monoclonal antibodies
- Removal of leached protein A, aggregates, host cell proteins, nucleic acids, and viruses from monoclonal antibodies is performed in flowthrough mode at which the antibodies pass directly through the column while the contaminants are adsorbed
- For optimal performance of Capto adhere, screening/optimization of loading conditions are required
- High capacity and productivity in flowthrough mode
- Savings in time and operating costs with a two-step chromatographic process
- Wide operational window of pH and conductivity
- Prepacked HiTrap columns for easy screening and convenient process development
Specifications
Specifications
| Diameter (Metric) | 7 mm |
| Flow Rate | <4 mL/min |
| For Use With | Removal of Contaminants to Formulation Levels in Post Protein A Purification |
| Functionality/Interaction Type | Strong Anion Exchanger with Multimodal Functionality |
| Length (Metric) | 25 mm |
| Ligand Type | N-benzyl methyl ethanolamine |
| Max. Pressure | 5 bar (70 psi) |
| Quantity | 1 Ea. |
| Content And Storage | Store at 4°C to 30°C, 20% Ethanol |
| Matrix | Capto |
| Show More |
Safety and Handling
Product Identifier
- HiTrap Capto adhere, 1 x 1 ml
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol
- UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction