Learn More
Cytiva HiTrap™ Benzamidine FF (high sub) Columns
Remove or purify serine proteases such as thrombin. Cytiva Life Sciences™ HiTrap™ Benzamidine FF (high sub) Columns are prepacked with Benzamidine Sepharose 4 Fast Flow (high sub).
Brand: Cytiva 17-5143-01
Code : GE
Additional Details : Weight : 2.14100kg
Description
- Prepacked with Benzamidine Sepharose™ 4 Fast Flow (high sub) media
- Protein binding capacity ≥35mg trypsin/mL medium
- Effective removal of thrombin and factor Xa after tag cleavage of recombinant proteins
Specifications
5 x 1mL | |
90 μm | |
1mL | |
0.7cm | |
0.98 in. | |
>12μM p-aminobenzamidine/mL medium | |
42psi | |
4% Highly Cross-linked Agarose | |
2° to 8°C |
<4 mL/min. | |
Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system | |
Benzamidine FF Column | |
Removal or purification of Serine Proteases such as Thrombin | |
2.5cm | |
Polypropylene | |
2 to 8 | |
Serine Proteases such as Thrombin |
Safety and Handling
- HiTrap Benzamidine FF (high sub) 1 ml, 5 x 1 ml
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P235-Keep cool.
- P501-Dispose of contents/container to .
Supplemental information
- MIXTURE LIST-Contains : ethanol