missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ AVB Sepharose™ Columns

Product Code. 11595784 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5 x 1 mL
Diameter (Metric) Inner:
7 mm
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Diameter (Metric) Inner unitSize
11595784 5 x 1 mL 7 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11595784 Supplier Cytiva Supplier No. 28411211

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Enables purification of adeno associated viruses (AAV) in one step, with high purity and excellent yields. Cytiva HiTrap™ AVB Sepharose™ Columns are pre-packed with affinity chromatography medium (resin) for simple operation with a syringe, pump or chromatographic system.

AVB Sepharose High Performance is an affinity resin designed for the purification of adeno associated virus (AAV). It enables purification of adeno associated viruses in one step with high purity and excellent yields.
  • One step purification of adeno associated viruses
  • Affinity resin with high selectivity and low background binding
  • Large surface area to give enhanced capacity
  • Prepacked HiTrap columns for simple operation with a syringe, pump, or chromatographic system
AVB Sepharose High Performance has proven affinity for adeno associated viruses from subclasses 1, 2, 3, and 5.
  • It is expected to have affinity for most of the other adeno associated virus subclasses
  • The ligand is a 14 kD fragment from a single chain antibody, expressed in yeast
TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 x 1 mL
Stability Stable to all commonly used aqueous buffers
For Use With (Application) One Step Purification of Adeno Associated Viruses
Storage Requirements Store prepacked column at 4°C to 30°C in 20% Ethanol and Media at 4°C to 8°C in 20% Ethanol.
Binding Capacity >10▿12 genecopies/mL Chromatography Mediμm
Product Type HiTrap AVB Sepharose HP Column
Diameter (Metric) Inner 7 mm
Flow Rate <150 cm/hr
Particle Size 24 to 44 μm
Height (Metric) Bed 25 mm
Material Polypropylene
Max. Pressure 70 psi (5 bar)
Matrix Cross-linked agarose
pH Range 3 to 10
Bed Volume (Metric) 1 mL
Ligand Type 14 kD recombinant protein
Show More Show Less
compliance-icons
Product Identifier
  • HiTrap AVB Sepharose High Performance 1 ml,5 x 1 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contain: ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.